ContactUsLinkComponent
Newton’s Color Disc
  470148-974
undefined
Newton’s Color Disc
  470148-974
 :  PH30701A
  • Description:
    Newton’s Color Disc
  • Cat. no.:
    470148-974
  • Supplier No.:
    PH30701A
What Happens When You Combine All Colors of Light?
  • Great classroom visual
  • Ideal for Elementary and Middle School levels
Easily demonstrate that all the colors of the visible spectrum can be combined to form white light with this simple device. Comprised of a pulley mounted on a metal stand, the seven colored metal disc is driven by a belt connected to a hand-driven wheel.