Neuromuscular Spindle Slide
470182-698
: Avantor
- Description:Neuromuscular Spindle Slide
- Cat. no.:470182-698
- Stain:Gold chloride
- Supplier No.:WARD470182-698
- Orientation:Whole mount
Helps brain determine body positioning
- Identifying features clearly distinguishable
- Rigorous quality control standards
Reptile. Sensory end-organ in muscle. Preparation of gold chloride impregnated muscle fibers.