Nervous Tissue, Central, Thin Section Slide
Nervous Tissue, Central, Thin Section, Thin section, Hematoxylin and Eosin
Catalog # 470183-670
Supplier: Avantor
Jump to:
Product Details & Documents
Extremely Thin Plastic Embedded Tissue For Greater Viewing Clarity
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Gray and white matter of spinal cord is demonstrated.
Spécifications
- Désignation:Nervous Tissue, Central, Thin Section
- Stain:Hematoxylin and Eosin
- Orientation:Thin section