Nervous Tissue, Central, Mammal Slide
Catalog # 470177-126
Supplier: Avantor
Specifications
- Description:Nervous Tissue, Central, Spinal cord Mammal Slide
- Cat. no.:470177-126
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470177-126
- Orientation:Cross section
Specifications
About this item
Illustrates mammalian nervous tissue
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Gray and white matter of spinal cord is demonstrated.