ContactUsLinkComponent
Nereis, Parapodium Slide
Catalog # 470181-586
Supplier:  Avantor
undefined
Nereis, Parapodium Slide
Catalog # 470181-586
Supplier:  Avantor
Specifications
  • Description:
    Nereis, Parapodium
  • Cat. no.:
    470181-586
  • Stain:
    Schneider's
  • Supplier No.:
    WARD470181-586
  • Orientation:
    Whole mount
Specifications
About this item
Polychaete Worm
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Structure used for respiration and locomotion, depending on the species.