Multi Purpose Fire Extinguishers | Avantor
Multi Purpose Fire Extinguishers
Supplier:   Logistics Supply
Multi Purpose Fire Extinguishers
Supplier:  Logistics Supply
Certificates
Multi Purpose Fire Extinguishers
2 Options Available Below
Product Details & Documents
For use on Fire Types (Classes) A, B, and C.
  • Pressure gauge provides at-a-glance status
  • Easy-to-pull safety pin
  • Powder coated cylinder for corrosion protection
  • 6-year limited warranty
These ideal classroom fire extinguishers are effective against all three types of fire; wood and paper, liquids and grease, and electrical. Both extinguishers contain dry chemical powder, pressurized with carbon dioxide gas. The powder is discharged in a blanket-like stream at a range of more than four meters to smother the fire. Both extinguishers are supplied with wall-mounting hardware.
 : Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Product Family Options
Informations sur le produit
  • Désignation
    UL Rating
    Availability
    Price
  • Multi-purpose fire extinguisher, 2.5 lbs.
    1A10B:C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $148.30
    $148.30
    Each
       $148.30
    Supplier Number: 21032136K
  • Multi-purpose fire extinguisher, 10 lbs.
    4A60B:C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $243.40
    $243.40
    Each
       $243.40
    Supplier Number: 466204K