Multi Colored Pipette Filler
Catalog # 470190-848
Supplier: Fischer Technical
Specifications
- Capacity:50 ml
- Description:Pipette Filler, Multi-Colored
- Cat. no.:470190-848
- Supplier No.:MULTIPIP
Specifications
About this item
Transfer corrosive, toxic or sterile liquids safely with this pipette filler. Three separate labeled valves make filling and emptying the pipette a simple procedure.