Porcelain Mortars
Fournisseur: SILICO & CHEMICO PORCELAIN SE
Certificats
À propos de cet article
High quality shallow mortars with pour-out lips.
- Autoclavable
The outside of each mortar is glazed; the inside of the mortar is unglazed. Pestles are sold separately.
Frequently Bought Together