Moray Eel
Moral Eel, Preserved, Vacuum-Pack
  470001-492
 :  134.1098.1
Moray Eel
  470001-492
Moray Eel
Moray Eel
Moral Eel, Preserved, Vacuum-Pack
$33.10
Each
Formalin Preserved
Gymnothorax miliaris. This slippery saltwater predator lurks inside reef crevaces in dark depths of water waiting for prey. Fascinating for dissections, this eel specimen features a second set of jaws, called pharyngeal jaws, that can extend to help in the capture of swallowing of prey. Size: 12–24".
  • Désignation:
    Moral Eel, Preserved, Vacuum-Pack
Accessories
  •  
    Description
    Grade
    Size
  • 470233-546
    WARDSafe™
    Laboratory
    19 L
    Each (19l) - $345.85