Mohr Buret Pinchcock Attachment
Catalogue # 470161-250
Fournisseur: Avantor
Spécifications
- Description:Pinchcock Attachment for Mohr Burets, for 60800 Series
- Cat. no.:470161-250
- Supplier No.:WARD470161-250
Spécifications
À propos de cet article
Designed for the Flint Glass Mohr Burets. Consists of a short piece of rubber tubing, glass buret tip, and a pinchcock clamp. Easily slips over the end of any buret not having a stopcock.