Modeling Clay
Modeling Clay: Red, Green, Blue, Yellow
470351-396
: X00740
Four individually wrapped colored sticks of easy-to-use Permoplast modeling clay can come in handy when doing setups for an optical bench. Each stick of clay weighs 0.25 lbs. Colors include red, yellow, blue, and green.
- Désignation:Modeling Clay: Red, Green, Blue, Yellow