Preserved Mink
Supplier: Ward's Science
Certificates
About this item
Formalin preserved mink. Available in vacuum packs or pails.
- Vertabrate anatomy
- Comparative anatomy
- Cat dissection substitute
Ranch-raised specimens are skinned, with all internal organs intact and undamaged. The feet may be cut and neck vertebrae broken due to processing, but this will not affect dissection and anatomy studies. Size: 14–18". Mink dissection makes a great alternative to cat dissection
Frequently Bought Together