Ward's® Mini Hovercraft Launcher
Mini hovercraft launcher
470360-228
New Product
Launch mini hovercrafts with different initial velocities.
- Removeable pegs allow for adjustable rubber band tension
- Accommodates hovercraft up to 20 cm (8")
This launcher allows you to vary the launch speed when releasing your pucks. Two separate post positions enable you to choose and alter the rate of motion for each launch. This wooden launching device features removeable rubber capped pegs and includes 10 rubber bands.
Designed for use with Air Puck, Kick It Stick It, and Power Soccer Discs, but accommodates any hovercraft up to 20 cm (8") in diameter.
- Désignation:Mini hovercraft launcher