Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Pre-cleaned slides
- L×W:3x1"
- Cat. no.:470151-068
- Supplier No.:630-004T
Quality microscope slides to make your own slide preparations.
- High quality slide making materials
- Dust free and ready to use
Pre-cleaned, premium grade microscope slides. Carefully selected to be non-corrosive and free of flaws, chemicals, and dust. Optically flat with ground edges and annealed to minimize chipping.