Kemtec® Microplates
Kemtec® Microplates
60, (30) 1.3 mL; (30) 3.8 mL, 127 mm, 76 mm
Catalog # 470199-952
Supplier:   Kemtec
Supplier #:  4-421
Kemtec® Microplates
Catalog # 470199-952
Supplier:  Kemtec
Kemtec® Microplates
Kemtec® Microplates
60, (30) 1.3 mL; (30) 3.8 mL, 127 mm, 76 mm
Price
$316.40
Log in for availability
Pack of 12 Case of 72
Product Details & Documents
These microplates replace standard test tubes and beakers. The dual-well plates are made of high-quality polystyrene and have 12 large wells and 48 small wells.
  • Closure type: slip-on lid
  • Base shape: flat base
  • Number of wells: 96
  • Max. volume: 390 µl
They are not for use with organic solvents.
Documents
Specifications
  • Nb de puits:
    60
  • Well Volume:
    (30) 1.3 mL; (30) 3.8 mL
  • Largeur:
    76 mm
  • Longueur:
    127 mm