Microdissection Forceps | Avantor
Microdissection Forceps
Supplier:   Dunrite Instruments
Microdissection Forceps
Certificates
Microdissection Forceps
2 Options Available Below
Product Details & Documents
Deep serrations in the jaws, plus aligning guide pins, make it easier to grasp and hold small objects. Choose from straight or curved points, both with a rounded tip.
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Tip Style
    Availability
    Price
  • Microdissection Forceps With Guide Pin
    Stainless steel
    101 mm (4")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7.20
    $7.20
    Each
       $7.20
    Supplier Number: 141420
  • Microdissection Forceps Without Guide Pin
    Stainless steel
    110 mm (4.5")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4.93
    $4.93
    Each
       $4.93
    Supplier Number: 141004