Mica Set
Mica Set
Catalog # 470024-862
Supplier: Avantor
Jump to:
Product Details & Documents
Specially selected thin sections of important rock forming minerals. Set may contain more than one example of the same mineral to depict variations in occurrence and distinct mineral associations.
Slides (12); annite, biotite, clinochlore, fuschite, glauconite, lepidolite, margarite, muscovite, paragonite, phlogopite, sericite, zinnwaldite.
Spécifications
- Désignation:Mica Set