Metridium Slide
Metridium, Cross section, Mallory Triple
470181-540
: Avantor
Plumose Sea Anemone
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shows typical arrangement of septa.
- Désignation:Metridium
- Stain:Mallory Triple
- Orientation:Cross section