Magnetizing and Demagnetizing Coil
Magnetizing and Demagnetizing Coil, 37×24 cm
Catalog # 470121-556
Supplier: Wards
Supplier #: PH0804B
Jump to:
Product Details & Documents
Bring older, weaker magnets back to life with this magnetizer.
- Coil allows magnets to be either remagnetized or demagnetized
A 25cm long coil allows magnets up to 4.5cm in diameter to be either remagnetized or demagnetized. The apparatus has 960 turns and the wire has a diameter of 1.22 mm. This coil serves as a useful way to demonstrate how a magnet can be made or destroyed. Two banana plug jacks allow for an AC/DC power supply to be attached, and an on/off switch is clearly visible.
Specifications
- Désignation:Magnetizing and Demagnetizing Coil
- Dimensions:37×24 cm