Ward's® Merismopedia | Avantor
Ward's® Merismopedia
Supplier:   Ward's Science
Ward's® <i>Merismopedia</i>
Supplier:  Ward's Science
Certificates
Ward's® <i>Merismopedia</i>
Ward's® Merismopedia
2 Options Available Below
Product Details & Documents
Freshwater. Colonial. Sheet of many cells one cell thick. Soil-water medium.
Shipped in a form that provides the best quality specimen. Most are supplied in 16 x 125 mm tubes, 2 oz. jars, or 8 oz. jars.
Product Family Options
Informations sur le produit
  • Désignation
    Media
    No. of Students
    Packaging
    Availability
    Price
  • Merismopedia Culture for 100 Students
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $47.30
    $47.30
    Each
       $47.30
    Supplier Number: -
  • Merismopedia (Small)
    Soil water medium
    30
    Tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $15.70
    $15.70
    Each
       $15.70
    Supplier Number: LB1902