Meiosis Microscope Slide Set
Catalogue # 470183-562
Fournisseur: Avantor
Spécifications
- Type:Cellular Development
- Description:Meiosis Microscope Slide Set
- No. of Slides:9
- Cat. no.:470183-562
- Supplier No.:WARD470183-562
Spécifications
À propos de cet article
Stages and Structures From a Lily Anther
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ordering information: Set includes Sporogenous tissue; Synizesis; Early prophase; Late prophase; First meiotic division; Second meiotic division; Pollen tetrads; Binucleate pollen; Single-celled microspores. All slides are prepared as cross-sections.