Medullated Nerve Fibers, Teased Preparation Slide
470182-690
: Avantor
- Description:Medullated Nerve Fibers, Teased Preparation Slide
- Cat. no.:470182-690
- Stain:Osmic Acid
- Supplier No.:WARD470182-690
- Orientation:Whole mount
Investigate nerve structures
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Peripheral nerves treated with osmic acid. Excellent display of nodes of Ranvier and medullary sheaths.