Media Bottles | Avantor
Media Bottles
Media Bottles
Certificates
Media Bottles
3 Options Available Below
Product Details & Documents
Media bottles are manufactured from borosilicate glass for excellent chemical resistance. A no-drop pour lip facilicates pouring and cleaning. Graduations show approximate volume, while a large writing patch can be used for identifying content.
Each bottle has a black phenolic PTFE rubber-lined cap attached. Caps are autoclavable.
Product Family Options
Informations sur le produit
  • Capacité
    Désignation
    Availability
    Price
  • 125 ml (4.2 oz.)
    Media Bottles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 48
    Case of 48 - $753.00
    $753.00
    Case of 48
       $753.00
    Supplier Number: 219815
  • 250 ml (8.5 oz.)
    Media Bottles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 48
    Case of 48 - $994.00
    $994.00
    Case of 48
       $994.00
    Supplier Number: 219817
  • 1 L (33.8 oz.)
    Media Bottles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $921.00
    $921.00
    Case of 24
       $921.00
    Supplier Number: 219820