Mechanical Engineering Models
Supplier:   Wards
Mechanical Engineering Models
Supplier:  Wards
Certificates
Mechanical Engineering Models
Mechanical Engineering Models
4 Options Available Below
Product Details & Documents
Clearly Observe Inner Workings
  • Demonstrate the most fundamental and most frequently used mechanisms with these mechanical engineering models
  • All components are precisely designed and can be assembled with clear sides to facilitate teaching
Useful for introducing engineering concepts, with models that are easy to assemble and disassemble. These models are great to visually demonstrate how the internal mechanisms of car transmissions, snowmobiles, elevators, and other mechanical items work. Students will be able to see things from how a simple transmission works with three forward gears and one reverse to the typical cone clutch drive used in many snowmobiles.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Worm Gear
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $77.60
    $77.60
    Each
       $77.60
  • Friction Wheel Drive
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $77.60
    $77.60
    Each
       $77.60
  • Cone Clutch Drive
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $77.60
    $77.60
    Each
       $77.60
  • Cam with Straight Line Lever
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $77.60
    $77.60
    Each
       $77.60
  • Three Speed and Reverse Gear
    Discontinued