Mayo Scissors | Avantor
Mayo Scissors
Supplier:   Dunrite Instruments
Mayo Scissors
Certificates
Mayo Scissors
2 Options Available Below
Product Details & Documents
Safe yet sharp.
  • Single blunt side prevents tissue damage
  • Available in 2 lengths
  • Stainless construction resists rust
Designed for use in the famous clinic, these straight-blade scissors allow for precise control. They are offered in two lengths. Stainless steel.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Blade Type
    Availability
    Price
  • Mayo Scissors
    140 mm (5.5")
    Stainless steel
    Sharp/Blunt
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $17.05
    $17.05
    Each
       $17.05
    Supplier Number: 141122
  • Mayo Scissors
    172 mm (6.75")
    Stainless steel
    Sharp/Blunt
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $22.60
    $22.60
    Log in pour la disponibilité
    Box of 10
    Box of 10 - $216.05
    $216.05
    Each
       $22.60
    Box of 10
       $216.05
    Supplier Number: 140992