Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Description:Making and Modeling Polymers Student Activity
- No. of Lab Groups:8
- Cat. no.:470039-462
- Includes:Teacher's Guide, Student Guides and Worksheets, SDS, Data Charts
- Supplier No.:706S
Spécifications
À propos de cet article
Introduce Students to the Structure and Properties of Polymer Molecules That Compose All Plastics
- Substances are often placed in categories or groups if they react or behave in similar ways
- Engineers design specific chemicals to be used in a variety of ways
- Models can be used to understand and explain scientific phenomena
- Requires 1-2 50 minutes classroom periods to complete
Investigations include exploring the linear polymer found in white glue, then adding a cross-linker to the glue and studying the resulting cross-linked polymer. Paper clip models are also used to discover how the molecular structural differences of monomers, polymers, and cross-linked polymers alter the properties of the molecules.