ContactUsLinkComponent
Magnetic Field Demonstrator–Iron Rod Type
Catalogue # 470016-024
Fournisseur:  Wards
undefined
Magnetic Field Demonstrator–Iron Rod Type
Catalogue # 470016-024
Fournisseur:  Wards
Supplier Number:  PH0802MF2
Spécifications
  • Description:
    Magnetic Field Demonstrator–Iron Rod Type
  • Cat. no.:
    470016-024
  • Supplier No.:
    PH0802MF2
Spécifications
À propos de cet article
A great visual for the classroom
  • Size: 26 x 26 x 1cm
  • The transparency of plates allows for overhead demonstrations
Plates with matrices of 98 small circular chambers contain an iron rod, which functions as effectively as a plotting compass. The transparent plates can be arranged in various ways including as faces of a cube or as a single large two-dimensional surface. This versatility allows you to show the reconfiguration of magnetic fields in one or two dimensions.