Longitudinal Wave Demonstration
Catalog # 470004-950
Supplier: Avantor
Specifications
- Description:Longitudinal Wave Demonstration
- Cat. no.:470004-950
- Supplier No.:WARD470004-950
Specifications
About this item
This inexpensive model, developed by Tim Ernest, lets you create longitudinal/compression waves by simply pushing a Slinky® along a track.
- Grades 6 to 9
- Length: 152.4 cm
It can be used in studies of waves, and sound to demonstrate compression, rarefaction, amplitude, wavelength, echoes, volume and pitch.
Delivery: Includes instructions.