Loligo Slide | Avantor
Loligo Slide
Loligo, Cross section, Hematoxylin and Eosin
Catalog # 470181-674
Supplier:   Avantor
<i>Loligo</i> Slide
Catalog # 470181-674
Supplier:  Avantor
<i>Loligo</i> Slide
Loligo Slide
Loligo, Cross section, Hematoxylin and Eosin
Price
$22.25
Log in pour la disponibilité
Each
Product Details & Documents
Demonstrates Myopsid Squid Body Type
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Squid. Shows body plan structure.
Spécifications
  • Désignation:
    Loligo
  • Stain:
    Hematoxylin and Eosin
  • Orientation:
    Cross section