Liquid Antiserums
Liquid Antiserums
Liquid Antiserums
Liquid Antiserums
Liquid antiserums are pooled and ready to use.
  • Anti-A and B Serums Identify ABO Blood Group
  • Anti-Rho (D) Identifies + or - Rh Factor
  • No Refrigeration Necessary
  • Pooled
  • Ready-To-Use
Anti-A and B serums identify ABO blood groups (A, B, AB, or O), and anti-AB provides a check on type O determinations. Anti-Rho (D) identifies + or - Rh factor.
Refrigeration is not necessary. The antiserums are contained in dropper bottles. They are for educational purposes only.
Product Family Options
  • Désignation
    No. of Students
  • Anti-Rho (D) 5 mL bottle
    50
    Each
    Each - $67.45
    $67.45
    Each
       $67.45
     : IS3152
  • Anti-A and Anti-B 
    50
    Each
    Each - $71.00
    $71.00
    Each
       $71.00
     : IS3150