Ward's® Limestone (Shell)
 :   Avantor
Ward's® Limestone (Shell)
 :  Avantor
Ward's® Limestone (Shell)
Ward's® Limestone (Shell)
Buff, coarse, crystalline “Austin Marble” with shell casts and molds.
  • Collected in Texas
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
  • Désignation
    Specimen Dimensions
  • Limestone (Shell), student specimens
    1×1 - 1×1"
    Pack of 10
    Pack of 10 - $13.85
    $13.85
    Pack of 10
       $13.85
  • Limestone (Shell), hand specimen
    2×2 - 3×4"
    Each
    Each - $32.05
    $32.05
    Each
       $32.05
  • Limestone (Shell), bulk pack, 1 kg
    Size will vary
    Each
    Each - $33.30
    $33.30
    Each
       $33.30
Storage
  •  
    Désignation
  • Storage box
    Each
    Each - $2.35
    $2.35
    Each
       $2.35