Ward's® Limestone (Encrinal)
Ward's® Limestone (Encrinal)
Supplier:   Avantor
Ward's® Limestone (Encrinal)
Supplier:  Avantor
Certificates
Ward's® Limestone (Encrinal)
Ward's® Limestone (Encrinal)
3 Options Available Below
Product Details & Documents
Gray with line crinoid columnal fragments.
  • Collected in Ohio
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kg of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range. Some crinoids may require a hand lens to observe.
Product Family Options
Specimens
Informations sur le produit
  • Désignation
    Specimen Dimensions
    Availability
    Price
  • Limestone (Encrinal), Student Specimens
    1×1 - 1×1"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $14.55
    $14.55
    Pack of 10
       $14.55
  • Limestone (Encrinal), Hand Specimen
    2×2" - 3×4"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $34.60
    $34.60
    Each
       $34.60
  • Limestone (Encrinal), Bulk Pack, 1 kg
    Size will vary
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $36.65
    $36.65
    Each
       $36.65
Storage
Informations sur le produit
  • Catalog #
    Désignation
    Availability
    Price
  • Storage box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2.35
    $2.35
    Each
       $2.35