Light Crystal Prism | Avantor
Light Crystal Prism
Light Crystal Prism
Catalog # 470007-090
Supplier:   Tedco
Fournisseur #:  00010
Light Crystal Prism
Catalog # 470007-090
Supplier:  Tedco
Light Crystal Prism
Light Crystal Prism
Light Crystal Prism
Price
$32.45
Log in pour la disponibilité
Each Pack of 6
Product Details & Documents
A unique light prism with a flat and slanted edge.
  • Uniquely shaped prism for optics experiments
  • Glass material
Split a beam of light into a multi-colored rainbow with the Light Crystal Prism!
Demonstrate a variety of optic experiments with the Light Crystal Prism - an equilateral prism with a special angled top. Use the enclosed instruction sheet for experiment ideas. The Light Crystal Prism measures approximately 60mm in height, with a 32mm face.
Spécifications
  • Désignation:
    Light Crystal Prism