Microcentrifuge Tube Lid Locks
Catalog # 470112-976
Supplier: United Scientific Supplies
Specifications
- Description:Microcentrifuge Tube Lid Locks, Mulit color with handle
- Cat. no.:470112-976
- Supplier No.:145063
Specifications
About this item
Secure caps to lids of microcentrifuge tubes with these autoclavable locks.
- Easy to use tabs
- Autoclavable
Each features a tab, allows to pick them out of a waterbath without burning your fingers.