Boreal Science Lens Cleaning Solution | Avantor
Boreal Science Lens Cleaning Solution
Supplier:   Ward's Science
Boreal Science Lens Cleaning Solution
Supplier:  Ward's Science
Certificates
Boreal Science Lens Cleaning Solution
Boreal Science Lens Cleaning Solution
1 Options Available Below
Product Details & Documents
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Lens Cleaning Solution
    100 ml
    Discontinued
    Supplier Number: MM0565-100
  • Lens Cleaning Solution
    100 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (100ml)
    Each (100ml) - $18.95
    $18.95
    Each (100ml)
       $18.95
    La quantité minimale de commande est de 1
    Supplier Number: MM0565-100ML