Ward's® Live Leeches
Ward's® Live Leeches
Ward's® Live Leeches
Certificates
Ward's® Live Leeches
Ward's® Live Leeches
5 Options Available Below
Product Details & Documents
Erpobdella obscura
  • Freshwater leeches
Typical examples of the class Hirudinea, these freshwater leeches are dorsoventrally flattened with prominent terminal suckers.
The species supplied may be blood suckers, predators, or scavengers.
Ordering information: These products contain living or perishable materials. Seasonal item, available from April to October, depending on weather.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Product Family Options
Informations sur le produit
  • Désignation
    Body Size
    Packaging
    Availability
    Price
  • Live leeches
    1 - 2"
    Pack of 1
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $20.45
    $20.45
    Each
       $20.45
    Supplier Number: 874720
  • Live leeches
    1 - 2"
    Pack of 3
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $48.93
    $48.93
    Each
       $48.93
    Supplier Number: -
  • Live leeches
    1 - 2"
    Pack of 6
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $56.80
    $56.80
    Pack of 6
       $56.80
    Supplier Number: 6616806
  • Live leeches
    1 - 2"
    Pack of 10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $74.20
    $74.20
    Pack of 10
       $74.20
    Supplier Number: 874721
  • Live leeches
    1 - 2"
    Pack of 12
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $85.25
    $85.25
    Each
       $85.25
    Supplier Number: -