Lead in Soil Test Kit | Avantor
Lead in Soil Test Kit
Lead in Soil Test Kit, 5
Catalog # 470173-836
Fournisseur #:  480311
Lead in Soil Test Kit
Catalog # 470173-836
CAS Number:  
Lead in Soil Test Kit
Lead in Soil Test Kit
Lead in Soil Test Kit, 5
CAS Number:  
Price
$68.60
Log in pour la disponibilité
Each Case of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Rare Test for Economical Price
  • Test for a common contaminant
  • Quick and simple to use
  • Instructions included
Simply make a slurry with your soil sample using the included reagents, then dip the slurry, and read the results. The detection level is 0–400 ppm. The kit includes instructions and enough materials for five tests.
Spécifications
Spécifications
  • Désignation:
    Lead in Soil Test Kit
  • No. of Tests:
    5