Dropping Bottles | Avantor
Dropping Bottles
Supplier:   Qorpak
Dropping Bottles
Supplier:  Qorpak
Certificates
Dropping Bottles
Dropping Bottles
3 Options Available Below
Product Details & Documents
Dropping bottles are made of low-density polyethylene.
  • White Polypropylene Dropper and Nasal Cap
  • Molded Plastic Screw Cap Seals at Tip
  • Low-Density Polyethylene
They feature a separable dropping tip.
The molded plastic screw cap seals at the tip.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Taille du bouchon
    Availability
    Price
  • 30 ml (1 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $109.50
    $109.50
    Pack of 12
       $109.50
    Supplier Number: 501182
  • 30 ml (1 oz.)
    18-400
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $107.90
    $107.90
    Case of 24
       $107.90
    Supplier Number: 501181
  • LDPE Dropping Bottles, Capacity: 60 mL
    60 ml (2 oz.)
    18 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $90.05
    $90.05
    Pack of 12
       $90.05
    Supplier Number: 501186