Latex Disposable Gloves
Latex Gloves, XL, White, Box/100
Catalog # 470150-774
Supplier: Dival Safety & Supplies
Fournisseur #: R10UNXL
Jump to:
Product Details & Documents
Cost effective hand protection.
- Ambidextrous
- Powdered
Disposable latex gloves reduce costs for hand protection where glove usage is high. Pre-powered for easy on and off. Perfect for your laboratory environment.
Spécifications
- Glove Color:White
- Taille du gant:XL
- Désignation:Latex Gloves
- Packaging:Box/100
- Powdered/Powder-free:Powder-free
- Cat. No.:470150-774
- Disposable / reusable:Disposable
- Ambidextrous/Hand-specific:Ambidextrous
- Latex/Latex-free:Latex