Large Dissecting Pan
Pan, Laboratory, Aluminum, w/o Wax, 33 cm x 24 cm x 6 cm
Catalog # 470008-088
Supplier: United Scientific Supplies
Fournisseur #: DSPA02
Jump to:
Product Details & Documents
Large aluminum dissecting pan.
- Easy to clean
- Rolled rim
Use this oversize pan to hold fetal pigs and other large specimens.
Constructed from durable, easy-to-clean heavy aluminum, the dissecting pan also offers a rolled rim for safety.
Spécifications
- Désignation:Pan, Laboratory, Aluminum, w/o Wax, 33 cm x 24 cm x 6 cm