LaMotte® Soil Texture Kit | Avantor
LaMotte® Soil Texture Kit
Supplier:   LaMotte
LaMotte® Soil Texture Kit
Supplier:  LaMotte
Certificates
LaMotte® Soil Texture Kit
LaMotte® Soil Texture Kit
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
With the Soil Texture Kit, students determine the percentage of sand, silt, and clay in soil samples.
  • Students Separate Different Sizes of Particles
  • For Use in the Field or the Lab
  • Instructions and Study Guide Included
  • Kit Includes Material for 50 Tests
  • Texture-Interpretation Guide Helps Students Decode Results
They can use the kit right in the field to see how sand, silt, and clay affect growth in the root zone.
The procedure allows them to separate the substances from the soil, determine their respective percentages in soil samples, and decode the results with an easy-to-read texture-interpretation guide.
Ordering information: This kit includes materials for 50 tests, a soil study guide, and instructions.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • LaMotte® Soil Texture Unit
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $158.80
    $158.80
    Each
       $158.80
    Supplier Number: 1067
  • Refill Kit for Soil Texture Kit, 50 tests
    Discontinued
    Supplier Number: R-1067