LaMotte® Bottom-Sampling Dredge
LaMotte® Bottom-Sampling Dredge, 67 cubic inches, 3", 7.5", 3"
Catalog # 470002-204
Supplier: LaMotte
Fournisseur #: 1097
Jump to:
Product Details & Documents
Designed for Sand or Silt
The trigger holds the sampler open during lowering in order to cover a uniform area, while the scissor design closes the stainless steel sampler when the trigger is released. A rope harness, available separately, attaches the arms to the line. Size: 7.5"L x 3"W x 3"D. Capacity: 67 cubic inches; weight: 7 lbs.
Spécifications
- Désignation:LaMotte® Bottom-Sampling Dredge
- Hauteur:3"
- Longueur:7.5"
- Nominal Capacity:67 cubic inches
- Width:3"