Laminaria Slide
Laminaria, Section, Quadruple
Catalog # 470181-012
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates the Multi-use Brown Algae (Kelp)
- Identifying features clearly distinguishable
- Rigorous quality control standards
Kelp blade with sporangia. This genus includes species that are edible and have medical applications as well.
Specifications
- Désignation:Laminaria
- Stain:Quadruple
- Orientation:Section