Telescoping Base Stand with Casters, 3'
Telescoping Base Stand with Casters
Catalog # 470203-156
Supplier: Labconco
Supplier #: 3730310
Jump to:
Product Details & Documents
These ADA-compliant base stands support up to 453.6kg (1000lbs.).
- Constructed of durable 4.5cm (1¾") tubular, glacier white epoxy-coated steel
- Stands have a depth of 73.7cm (29")
Stands with telescoping legs are adjustablle to seven height positions in 2.5cm (1') increments from 69.9 - 85.1cm (27½ - 33½").
Ordering information: Base stand only.
Specifications
- Désignation:Telescoping Base Stand with Casters