Lab Label Roll
Lab Label Roll
Lab Label Roll
Lab Label Roll
Better than gummed labels, these pressure-sensitive labels adhere securely, yet can be easily removed.
  • Autoclavable to 260 °C
The labels are on a continuous roll.
Product Family Options
  • Désignation
    Width
    Longueur
  • Lab label roll, dispenser
    ¾"
    100"
     : 130575
  • Lab label roll
    ¾"
    100"
    Each
    Each - $27.05
    $27.05
    Each
       $27.05
     : 15-1970