ContactUsLinkComponent
KIMAX® Gas Measuring Tubes
Catalog # 470235-788
Supplier:  GSC International
undefined
KIMAX® Gas Measuring Tubes
Catalog # 470235-788
Supplier:  GSC International
Supplier Number:  701-1-10
Specifications
  • Description:
    Kimax Gas Measuring Tubes, 50 x 0.1 mL
  • Cat. no.:
    470235-788
Specifications
About this item
Made from high quality borosilicate glass.
  • The top is closed with the tube graduated from the top down