ContactUsLinkComponent
KIMAX® Fermentation Tubes with Foot
Catalogue # 470104-226
Fournisseur:  GSC International
undefined
KIMAX® Fermentation Tubes with Foot
Catalogue # 470104-226
Fournisseur:  GSC International
Supplier Number:  601-9
Spécifications
  • Description:
    Kimax Fermentation Tubes with Foot, 100 mm x 13 mm
  • Cat. no.:
    470104-226
  • Supplier No.:
    601-9
Spécifications
À propos de cet article
For use in the collection and observation of gases produced by fermentation.
  • Made from borosilicate glass, the bulb is designed to have a capacity greater than that of the vertical tube
  • The vertical tube has a closed, rounded top