Specifications
- Description:K-9 Bandaging Manikin
- Cat. no.:470029-830
- Supplier No.:525
Specifications
About this item
Versatile Manikin For Advanced First Aid Procedures
- Full body manikin for practicing multiple techniques
- Made to order
- Includes carrying bag
This full-sized model is ideal for practical lab skills and practicing advanced bandaging techniques. Apart from administering first aid, the manikin can be used to practice mouth-to-snout resuscitation. The left knee, elbow, carpal and hock joints are capable of 90° movements. Shoulder and hip joints also have the flexibility of 90° rotation.
With the new features, users can now also practice the Velpeau Sling on the right forelimb and the Ehmer Sling on the right hind leg.
In short, these are just some of the bandaging techniques users can practice on Rufus:
90-90- flexion sling
Carpal sling
Robert Jones bandage
Velpeau sling
Ehmer sling
Limb bandage using stirrups
Trunk bandage
Tail bandage
Padded hip bandage
Head and Ear bandage
Casts
Rufus comes with five disposable lungs, a brush, and carrying case with kneeling pad.