Jumbo Wall Clock | Avantor
Jumbo Wall Clock
Jumbo wall clock, 17.1×27.9×5.1 cm
Catalog # 470014-816
Fournisseur #:  CL030025
Jumbo Wall Clock
Catalog # 470014-816
Jumbo Wall Clock
Jumbo Wall Clock
Jumbo wall clock, 17.1×27.9×5.1 cm
Price
$301.80
Log in pour la disponibilité
Each
Product Details & Documents
The Jumbo wall clock is easily read from 30 feet away.
  • Six time zones
  • Digital
  • Clock displays time, date and temperature
  • NIST radio signal automatically adjusts clock
  • Celsius or Fahrenheit temperature readings
This large digital clock covers six time zones and simultaneously displays the time, date, and temperature. A controlling radio signal from NIST in Boulder, Colorado automatically adjusts the clock to its precise time, which can be shown as 12-hour AM/PM time or 24-hour international time. A calendar displays the month, date, and day of the week, and the clock has Celsius or Fahrenheit temperature readings.
Additional features include a daily alarm and snooze function and a low-battery indicator.
Delivery information: This clock includes two built-in table stands and three required C batteries.
Spécifications
  • Désignation:
    Jumbo wall clock
  • W×H×D:
    17.1×27.9×5.1 cm