Iris Scissors | Avantor
Iris Scissors
Supplier:   Dunrite Instruments
Iris Scissors
Certificates
Iris Scissors
2 Options Available Below
Product Details & Documents
Extra control in tight spaces.
  • Fine point scissors for fine cutting
  • Sharp tips give better control
  • Available in straight or angled blades
Choose from straight or angled scissors, both with sharp points to facilitate finer dissection work. The angled style is often preferred for hard-to-reach tissues. First quality stainless steel.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Blade Type
    Availability
    Price
  • Iris Scissors, Straight
    110 mm (4.5")
    Stainless steel
    Sharp/Sharp
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $14.40
    $14.40
    Each
       $14.40
    Supplier Number: 140611
  • Iris Scissors, Angled
    110 mm (4.5")
    Stainless steel
    Sharp/Sharp
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $12.50
    $12.50
    Each
       $12.50
    Supplier Number: 140975